Learn More
Stoelting™ Mosquito Forceps
SDP
Catalog No. 10001358 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Curved, Delicate
Straight, Delicate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-001-358 Curved, Delicate
10-001-357 Straight, Delicate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-001-358 Supplier Stoelting™ Supplier No. 5211040
Add to Cart
Add to Cart
Hemostatic forceps are used in many surgical procedures usually to control bleeding or the flow of liquids in tubing.
  • Scissors-like ring handles
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
  • Delicate hemostats have thin jaws with fine serrations.
Specifications
For Use With (Application) Neuroscience
Length (English) 8.25 in.
Model Curved, Delicate