Learn More
Sklar Instruments Hartmann Mosquito Forceps, Sklar, 3-1/2 in., 1x2 Teeth, Curved, Serrated, Stainless Steel. Sklar Hartmann Mosquito forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures. EA/1.
SDP
Catalog No. 501437394 Shop All Sklar Instruments Products
Encompass_Preferred
Hartmann Mosquito Forceps, Sklar, 3-1/2 in., 1x2 Teeth, Curved, Serrated, Stainless Steel. Sklar Hartmann Mosquito forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures. Most commonly utilized in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures. Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps. Sklar operating room grade instruments are made from the highest grade surgical steel and incorporate the finest level of craftsmanship. Superior manufacturing and quality inspection guarantee that Sklar instruments meet the demands of our customers.
This item is not returnable. View return policy
Catalog No. 50-143-7394 Supplier Sklar Instruments Supplier No. 171336
Hartmann Mosquito Forceps, Sklar, 3-1/2 in., 1x2 Teeth, Curved, Serrated, Stainless Steel. Sklar Hartmann Mosquito forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures. Most commonly utilized in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures. Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps. Sklar operating room grade instruments are made from the highest grade surgical steel and incorporate the finest level of craftsmanship. Superior manufacturing and quality inspection guarantee that Sklar instruments meet the demands of our customers.