Learn More
Sklar Instruments Econo Hartmann Mosquito Forceps, Sklar, 3-1/2 in., Straight, Serrated, Stainless Steel. PK/12.
SDP
Catalog No. 501437634 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Hartmann Mosquito Forceps, Sklar, 3-1/2 in., Straight, Serrated, Stainless Steel. Econo Hartmann Mosquito Forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures. Most commonly utilized in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures. Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 3-1/2 inches. They are sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7634 Supplier Sklar Instruments Supplier No. 21409
Econo Hartmann Mosquito Forceps, Sklar, 3-1/2 in., Straight, Serrated, Stainless Steel. Econo Hartmann Mosquito Forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures. Most commonly utilized in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures. Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 3-1/2 inches. They are sold in boxes of 12.