Learn More
World Precision Instrument Mosquito Hemostatic Forceps, Straight, 9.5cm, Titanium
SDP
Catalog No. NC1956359
Encompass
Mosquito Forceps, also called mosquito hemostatic forceps or simply hemostats, are surgical instruments designed for clamping blood vessels or other small tissues to control bleeding during surgery or to provide a secure grip. It was thought that their long, slender tips resemble the legs of a mosquito. Life science researchers like these fine hemostats because of the secure grip and the ability to manipulate them in confined spaces. Our Titanium Mosquito Forceps have long, thin, and straight jaws that taper to a fine tip. The jaws are serrated for a firm grasp with minimal tissue trauma. The handles of Mosquito Forceps have a ratchet lock to secure your grasp once it is engaged. These are available in 12 cm (4.7") long. 
Catalog No. NC1956359 Supplier World Precision Instrument Supplier No. WP509900
Mosquito Forceps, also called mosquito hemostatic forceps or simply hemostats, are surgical instruments designed for clamping blood vessels or other small tissues to control bleeding during surgery or to provide a secure grip. It was thought that their long, slender tips resemble the legs of a mosquito. Life science researchers like these fine hemostats because of the secure grip and the ability to manipulate them in confined spaces. Our Titanium Mosquito Forceps have long, thin, and straight jaws that taper to a fine tip. The jaws are serrated for a firm grasp with minimal tissue trauma. The handles of Mosquito Forceps have a ratchet lock to secure your grasp once it is engaged. These are available in 12 cm (4.7") long. 