Learn More
Anton Paar Most advanced four-digit density meter in its price classConvenient filling via Xsample 200Two syringe filling positions for user-friendly operationConsistent results with 0.00005 g/cm3 repeatabilityCoverage of ASTM, ISO, and Pharmacopoeia standards
Catalog No. 5000580481
Encompass_Preferred
SPECIFICATIONSMeasuring range:Density: 0 g/cm³ to 3 g/cm³Temperature: 15°C to 60 °C (59 °F to 140 °F)*Accuracy:Density: 0.0001 g/cm³ Temperature: 0.03 °C (0.02 °F)Repeatability, s.d.Density: 0.00005 g/cm³Temperature: 0.02 °C (0.04 °F)Reproducibility, s.d.Density: 0.00007 g/cm³Digital ResolutionTemperature: 0.01 °C (0.01 °F)
Catalog No. 50-005-80481 Supplier Anton Paar Supplier No. 326145
SPECIFICATIONSMeasuring range:Density: 0 g/cm³ to 3 g/cm³Temperature: 15°C to 60 °C (59 °F to 140 °F)*Accuracy:Density: 0.0001 g/cm³ Temperature: 0.03 °C (0.02 °F)Repeatability, s.d.Density: 0.00005 g/cm³Temperature: 0.02 °C (0.04 °F)Reproducibility, s.d.Density: 0.00007 g/cm³Digital ResolutionTemperature: 0.01 °C (0.01 °F)