Learn More
Bachem Fmoc-OSu
SDP
Catalog No. 501947387
Encompass_Preferred
Fmoc-OSu, 82911-69-1, C19H15NO5, Mr 337.33, 25g. N-(9-Fluorenylmethyloxycarbonyloxy)-succinimide. Most efficient reagent for the selective preparation of Fmoc-derivatives of hydroxy amino acids in high yield. The advantage of Fmoc-OSu is that little or no oligopeptides are formed when amino acid derivatives are prepared. The use of this reagent in the synthesis of glycopeptides has been reported as well.
Catalog No. 50-194-7387 Supplier Bachem Supplier No. 4006805.0025
Fmoc-OSu, 82911-69-1, C19H15NO5, Mr 337.33, 25g. N-(9-Fluorenylmethyloxycarbonyloxy)-succinimide. Most efficient reagent for the selective preparation of Fmoc-derivatives of hydroxy amino acids in high yield. The advantage of Fmoc-OSu is that little or no oligopeptides are formed when amino acid derivatives are prepared. The use of this reagent in the synthesis of glycopeptides has been reported as well.