Learn More
Anton Paar Most powerful rheometer for special applicationsMax torque (300 mNm) and normal force (70 N) specificationsEnables long-term measurements at high torque
Catalog No. 5000580447
Encompass_Preferred
MCR 503 Power SpecificationsBearing design: Air, fine-pored carbonMotor design: Electronically commutated (EC) permanent magnet synchronous motorDisplacement transducer: High-resolution optical encoderNormal force measurement design: 360° capacitive sensor, non-contacting, fully integrated in bearingWorking Modules: CMTMin. torque (rotation): 100 nNmMin. torque (oscillation): 50 nNmMax. torque: 300 mNmTorque resolution: 0.2 nNmAngular deflection resolution: < 1 nradMin. angular velocity: 0 rad/sMax. angular velocity / max. speed: 200 rad/s (2,100 rpm)Min. frequency: 2 x 10-8 HzMax. frequency: 200 HzNormal force range: 0.001 N to 70 NNormal force resolution: 0.1 mNTruStrain: YesReady for DMA in tension, bending, and compression: Yes
Catalog No. 50-005-80447 Supplier Anton Paar Supplier No. 224625
Please contact your sales representative for a quote or further ordering instructions.
MCR 503 Power SpecificationsBearing design: Air, fine-pored carbonMotor design: Electronically commutated (EC) permanent magnet synchronous motorDisplacement transducer: High-resolution optical encoderNormal force measurement design: 360° capacitive sensor, non-contacting, fully integrated in bearingWorking Modules: CMTMin. torque (rotation): 100 nNmMin. torque (oscillation): 50 nNmMax. torque: 300 mNmTorque resolution: 0.2 nNmAngular deflection resolution: < 1 nradMin. angular velocity: 0 rad/sMax. angular velocity / max. speed: 200 rad/s (2,100 rpm)Min. frequency: 2 x 10-8 HzMax. frequency: 200 HzNormal force range: 0.001 N to 70 NNormal force resolution: 0.1 mNTruStrain: YesReady for DMA in tension, bending, and compression: Yes