Learn More
Selleck Chemical LLC Motesanib
SDP
Catalog No. 5000000080
Encompass_Preferred
Motesanib (AMG-706) is an orally bioavailable receptor tyrosine kinase inhibitor with IC50 values of 2 nM 3 nM 6 nM 8 nM 84 nM 59 nM for VEGFR1 VEGFR2 VEGFR3 Kit PDGFR and Ret respectively
Catalog No. 50-000-00080 Supplier Selleck Chemical LLC Supplier No. S579310MG
Motesanib (AMG-706) is an orally bioavailable receptor tyrosine kinase inhibitor with IC50 values of 2 nM 3 nM 6 nM 8 nM 84 nM 59 nM for VEGFR1 VEGFR2 VEGFR3 Kit PDGFR and Ret respectively