Learn More
Enzo Life Sciences Motesanib (5mg). CAS: 453562-69-1
SDP
Catalog No. 502013451
Encompass_Preferred
Motesanib potently inhibits angiogenesis and induces regression in tumor xenografts. Alternative name: AMG-706. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or ethanol. Long Term Storage: -80°C.
Catalog No. 50-201-3451 Supplier Enzo Life Sciences Supplier No. ENZCHM1470005
Motesanib potently inhibits angiogenesis and induces regression in tumor xenografts. Alternative name: AMG-706. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or ethanol. Long Term Storage: -80°C.