Learn More
TARGETMOL CHEMICALS INC Motesanib Diphosphate 50MG
SDP
Catalog No. 502877716
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Motesanib Diphosphate (AMG 706) is the orally bioavailable diphosphate salt of a multiple-receptor tyrosine kinase inhibitor with potential antineoplastic activity. Purity 99.76%
Catalog No. 50-287-7716 Supplier TARGETMOL CHEMICALS INC Supplier No. T2288L50MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Motesanib Diphosphate (AMG 706) is the orally bioavailable diphosphate salt of a multiple-receptor tyrosine kinase inhibitor with potential antineoplastic activity. Purity 99.76%