Learn More
Medchemexpress LLC Motesanib diphosphate | 857876-30-3 | MFCD12407403 | >98.0% | 569.44 | C22H29N5O9P2 | 50 MG
SDP
Catalog No. 5000246383
Encompass_Preferred
Motesanib diphosphate is the diphosphate salt of motesanib, an ATP-competitive small-molecule inhibitor of VEGF receptors used as a research reagent for kinase and signaling studies.
  • ATP-competitive inhibitor of VEGFR1/2/3 with low-nanomolar potency.
  • Diphosphate salt form improves aqueous solubility for in vitro applications.
  • Molecular weight 569.44; formula C22H29N5O9P2.
  • High solubility in DMSO (≥110 mg/mL) and moderate solubility in water (≈5 mg/mL with warming).
  • Intended for research and analytical use; not for human or clinical use.
Catalog No. 50-002-46383 Supplier Medchemexpress LLC Supplier No. HY1022950MG
Motesanib diphosphate is the diphosphate salt of motesanib, an ATP-competitive small-molecule inhibitor of VEGF receptors used as a research reagent for kinase and signaling studies.
  • ATP-competitive inhibitor of VEGFR1/2/3 with low-nanomolar potency.
  • Diphosphate salt form improves aqueous solubility for in vitro applications.
  • Molecular weight 569.44; formula C22H29N5O9P2.
  • High solubility in DMSO (≥110 mg/mL) and moderate solubility in water (≈5 mg/mL with warming).
  • Intended for research and analytical use; not for human or clinical use.