Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Motesanib diphosphate | 857876-30-3 | MFCD12407403 | 99.9% | 569.44 | C22H29N5O9P2 | 25 MG
SDP

Catalog No. 5000246744
Encompass_Preferred

Motesanib diphosphate is a small-molecule, ATP-competitive inhibitor of vascular endothelial growth factor receptors VEGFR1, VEGFR2, and VEGFR3 that also inhibits Kit. The diphosphate salt is supplied for research and analytical applications.

  • Potent VEGFR1/2/3 inhibition (IC50 ~2 nM, 3 nM, 6 nM).
  • Also inhibits Kit, with ~10-fold selectivity for VEGFR versus PDGFR and Ret.
  • High purity (≈99.86%) with molecular weight 569.44 and formula C22H29N5O9P2.
  • Supplied as a 25 mg research/analytical sample.
  • Intended for non-clinical, research use only.

Catalog No. 50-002-46744 Supplier Medchemexpress LLC Supplier No. HY1022925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Motesanib diphosphate is a small-molecule, ATP-competitive inhibitor of vascular endothelial growth factor receptors VEGFR1, VEGFR2, and VEGFR3 that also inhibits Kit. The diphosphate salt is supplied for research and analytical applications.

  • Potent VEGFR1/2/3 inhibition (IC50 ~2 nM, 3 nM, 6 nM).
  • Also inhibits Kit, with ~10-fold selectivity for VEGFR versus PDGFR and Ret.
  • High purity (≈99.86%) with molecular weight 569.44 and formula C22H29N5O9P2.
  • Supplied as a 25 mg research/analytical sample.
  • Intended for non-clinical, research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.