Learn More
Medchemexpress LLC Motesanib diphosphate | 857876-30-3 | MFCD12407403 | 99.9% | 569.44 g/mol | C22H29N5O9P2 | 1 ML
SDP
Catalog No. 5000246385
Encompass_Preferred
Motesanib diphosphate is the diphosphate salt of motesanib (AMG 706), a potent ATP-competitive inhibitor of VEGFR1/2/3. It is supplied for research and analytical use to prepare concentrated stock solutions and for use as a reference standard in biochemical and cell-based assays.
  • Research reagent for kinase inhibition assays.
  • High purity, 99.9% (reported by supplier).
  • CAS number 857876-30-3 and molecular weight 569.44 g/mol.
  • Provided to support preparation of 1, 5, and 10 mM stock solutions.
  • Common pack sizes listed for preparation include 1 mg, 5 mg, and 10 mg.
Catalog No. 50-002-46385 Supplier Medchemexpress LLC Supplier No. HY1022910MM 1ML
Motesanib diphosphate is the diphosphate salt of motesanib (AMG 706), a potent ATP-competitive inhibitor of VEGFR1/2/3. It is supplied for research and analytical use to prepare concentrated stock solutions and for use as a reference standard in biochemical and cell-based assays.
  • Research reagent for kinase inhibition assays.
  • High purity, 99.9% (reported by supplier).
  • CAS number 857876-30-3 and molecular weight 569.44 g/mol.
  • Provided to support preparation of 1, 5, and 10 mM stock solutions.
  • Common pack sizes listed for preparation include 1 mg, 5 mg, and 10 mg.