Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Motesanib diphosphate | 857876-30-3 | MFCD12407403 | 99.9% | 569.44 g/mol | C22H29N5O9P2 | 100 MG
SDP

Catalog No. 5000245856
Encompass_Preferred

Motesanib diphosphate is the diphosphate salt of a small-molecule ATP-competitive inhibitor of vascular endothelial growth factor receptors (VEGFR1/2/3). It is supplied as a research-grade analytical standard for biochemical assays, pathway studies, and method development. The compound should be stored sealed and protected from moisture; in solvent, stability is -80°C for up to six months and -20°C for up to one month.

  • High purity (~99.9%).
  • Potent VEGFR1/2/3 inhibition with IC50s 2-6 nM.
  • Molecular formula C22H29N5O9P2, molecular weight 569.44 g/mol.
  • Suitable for biochemical assays and analytical method development.
  • Storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-45856 Supplier Medchemexpress LLC Supplier No. HY10229100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Motesanib diphosphate is the diphosphate salt of a small-molecule ATP-competitive inhibitor of vascular endothelial growth factor receptors (VEGFR1/2/3). It is supplied as a research-grade analytical standard for biochemical assays, pathway studies, and method development. The compound should be stored sealed and protected from moisture; in solvent, stability is -80°C for up to six months and -20°C for up to one month.

  • High purity (~99.9%).
  • Potent VEGFR1/2/3 inhibition with IC50s 2-6 nM.
  • Molecular formula C22H29N5O9P2, molecular weight 569.44 g/mol.
  • Suitable for biochemical assays and analytical method development.
  • Storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.