Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Motesanib diphosphate | 857876-30-3 | MFCD12407403 | 99.9% | 569.44 g/mol | C22H29N5O9P2 | 5 MG
SDP

Catalog No. 5000245855
Encompass_Preferred

Motesanib diphosphate is the diphosphate salt of motesanib, an ATP-competitive inhibitor of VEGFR1/2/3 and c-Kit, supplied for laboratory research and analytical applications.

  • Potent ATP-competitive inhibitor of VEGFR1/2/3 and c-Kit.
  • Molecular formula: C22H29N5O9P2.
  • Molecular weight: 569.44 g/mol.
  • Pack size: 5 mg.
  • Solubility: DMSO ≥ 110 mg/mL; H2O 5 mg/mL (with ultrasonic warming).
  • Storage: 4°C sealed; in solvent -80°C (6 months), -20°C (1 month).
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-45855 Supplier Medchemexpress LLC Supplier No. HY102295MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Motesanib diphosphate is the diphosphate salt of motesanib, an ATP-competitive inhibitor of VEGFR1/2/3 and c-Kit, supplied for laboratory research and analytical applications.

  • Potent ATP-competitive inhibitor of VEGFR1/2/3 and c-Kit.
  • Molecular formula: C22H29N5O9P2.
  • Molecular weight: 569.44 g/mol.
  • Pack size: 5 mg.
  • Solubility: DMSO ≥ 110 mg/mL; H2O 5 mg/mL (with ultrasonic warming).
  • Storage: 4°C sealed; in solvent -80°C (6 months), -20°C (1 month).
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.