Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Motic™ Moticam 4000X

Catalog No. 01001647 Shop All Motic Group Products
Change view
Click to view available options
For Use With (Equipment):
Motic Trinocular Microscopes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-001-647 Motic Trinocular Microscopes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-647 Supplier Motic™ Supplier No. 1100600101641
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Moticam 4000X is a Full HD affordable multi-tasking microscopy camera with True 4K imaging.

  • Discover our newest Moticam 4000X, designed for efficiency and to output fast live images at 4K (3840 x 2160 px).
  • Connect it through the HDMI port to a screen and display your live image. With the use of the on-board software and a USB mouse, you can make live measurements, notes, image adjustments and more, directly on the HDMI screen, without the need for a computer.
  • Thanks to its integrated SD card slot and USB ports, saving images won't require a computer, making it easy to save and organize your images and videos. You can also connect your camera through USB to a Windows, or Mac computer, and even display your samples simultaneously.

Specifications

Type Digital HDMI Camera
CAS 67-56-1
Autoclavable Not Autoclavable
Camera Type HDMI Camera for Microscope
Color Black
Linear Formula C12H14D5NO · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
For Use With (Equipment) Motic Trinocular Microscopes
Formula Weight 234.78
Includes CS ring adaptor, focusable lens, 30mm and 38mm eyepiece adapters, Motic 4-dot calibration slide, macro calibration card, macro tube, universal power supply, HDMI cable, USB 3.0 cable, Motic Images Plus 3.1
Packaging Ampule
Pixels 1.45 x 1.45 μm
Port Type HDMI, USB, Wi-Fi, Ethernet, Micro SD-card
Resolution 8MP (3840 x 2160 pixels)
Sensor Types CMOS
Size 1/2.8 in.
Sterility Non-sterile
Voltage 12 V
Frequency (Frame Rate) HDMI: 4K (3840x2160) at 30fps USB 3.0: 1920x1080 at 30fps, 3840x2160 at 25fps Wi-Fi: 1920x1080 at25fps, 3840x2160 at 12fps
Quantity 1 mL
Flash Point 9.7°C (49.5°F)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.