Learn More
Motic™ Moticam A1
Catalog No. 01258033 Shop All Motic Group Products
Change view
Click to view available options
Sensor Types:
sCMOS
Catalog No. Sensor Types
01-258-033 sCMOS
1 options
This item is not returnable. View return policy
Catalog No. 01-258-033 Supplier Motic™ Supplier No. 1100600101412
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Moticam A1 is ideal for newcomers and advanced enthusiasts who are looking for an affordable digital microscope instrument. Capture everything you need at 720p and work at a steady frame rate with all accessories included.
  • C-mount for lens
  • MotiConnect software is available for free download (with free updates included)
  • Live Display Modes: 1280 x 720 px, 1024 x 768 px
Specifications
Type Camera
Product Line Moticam A
Includes Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap
Sensor Types sCMOS
Size 1/3.1 in.
Frequency (Frame Rate) 30 fps
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov