Learn More
Motic™ Moticam A16
Catalog No. 01258037 Shop All Motic Group Products
Change view
Click to view available options
Sensor Types:
sCMOS
Catalog No. Sensor Types
01-258-037 sCMOS
1 options
This item is not returnable. View return policy
Catalog No. 01-258-037 Supplier Motic™ Supplier No. 1100600101463
Add to Cart
Add to Cart
This item is not returnable. View return policy
The ultimate Moticam for advanced enthusiasts with demanding needs, the Moticam A16 has a large 1/2 in. sCMOS sensor with resolution above 4K and delivers up to 15 fps in live imaging.
  • C-mount for lens
  • MotiConnect software is available for free download (with free updates included)
  • Live Display Modes: 4608 x 3456 px, 3264 x 2448 px, 1920 x 1080 px
  • Maximum Frame Rate: 4608 x 3456 at 15 fps, 3264 x 2448 at 15 fps, 1920 x 1080 at 30 fps
Specifications
Type Camera
Product Line Moticam A
Includes Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap
Sensor Types sCMOS
Size 1/2.3 in.
Frequency (Frame Rate) 15 to 30 fps
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov