Learn More
Motic™ Moticam A8
Catalog No. 01258036 Shop All Motic Group Products
Change view
Click to view available options
Sensor Types:
sCMOS
Catalog No. Sensor Types
01-258-036 sCMOS
1 options
This item is not returnable. View return policy
Catalog No. 01-258-036 Supplier Motic™ Supplier No. 1100600101443
Add to Cart
Add to Cart
This item is not returnable. View return policy
The fast sCMOS imaging setup of the Moticam A8 allows 4K resolution while delivering steady 15 fps. Printing images requires high-resolution data; Moticam delivers the detail needed.
The ideal camera, for everyone who has high demands and wants to stay flexible.
  • C-mount for lens
  • MotiConnect software is available for free download (with free updates included)
  • Live Display Modes: 3840 x 2160 px, 1920 x 1080 px
  • Maximum Frame Rate: 3840 x 2160 at 15 fps, 1920 x 1080 at 30 fps
Specifications
Type Camera
Product Line Moticam A
Includes Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, Calibration Slide, Macro Viewing Tube, C-ring, USB Cable, Dust Cap
Sensor Types sCMOS
Size 1/3 in.
Frequency (Frame Rate) 15 to 30 fps
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov