Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Motic™ Moticam BMH4000X

Catalog No. 01001648 Shop All Motic Group Products
Change view
Click to view available options
Sensor Types:
CMOS
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sensor Types
01-001-648 CMOS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-648 Supplier Motic™ Supplier No. 1100600101731
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Moticam BMH4000X is a Full-HD C-Mount multitasking microscopy camera that delivers an impressive Full HD live image with maximum colour fidelity on an integrated LCD screen.

  • The new Moticam BMH4000X is a camera that was designed for ease of use and the ability to output fast live images at 4k(3840 x 2160) pixels in full HD.

  • Unlike other cameras, the Moticam BMH4000X can be connected to a computer or through HDMI. Simply plug the camera through the HDMI port and display images with the use of the onboard software and a USB mouse through the built-in 11.6 in. LCD screen, or connect your camera through USB to a Windows, Mac or Linux computer.

  • By having an integrated SD card slot, not only does saving images not require a computer, but it also allows flexibility to save portable data.

Specifications

Type Digital HDMI Camera
CAS 75-05-8, 928822-98-4
Autoclavable Not Autoclavable
Camera Type HDMI Camera for Microscope
Color Black
Linear Formula C17H22N2O
MDL Number MFCD11655942
UN Number UN 1648 3 / PGII
Concentration 1.0 mg/mL in acetonitrile
Formula Weight 270.37
Packaging Ampule
Pixels 1.45 x 1.45 μm
Resolution 8MP (3840 x 2160 pixels)
Sensor Types CMOS
Size 1/2.8 in.
Sterility Non-sterile
Voltage 12 V
Frequency (Frame Rate) HDMI: (4K) 30fps at 3840x2160 WIFI: 25fps at 1920x10180, 12fps at 3840x2160 USB: (4K) 30fps at 3840x2160
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.