Learn More
Motic™ Moticam S12 and Analysis Basic Software
Catalog No. 01257468 Shop All Motic Group Products
Change view
Click to view available options
Sensor Types:
sCMOS
Catalog No. Sensor Types
01-257-468 sCMOS
1 options
This item is not returnable. View return policy
Catalog No. 01-257-468 Supplier Motic™ Supplier No. 1100600101211
Add to Cart
Add to Cart
This item is not returnable. View return policy
For maximum resolution for printing purposes, Moticam S12 is your choice. Ideal for high resolution in display or for printing purposes.
Motic™'s new C-mount ready sCMOS cameras are the newest members of the Moticam family of professionally designed and engineered cameras. It is easy enough for the microscopy novice, but sophisticated enough for the professional. With back illuminated sensor technology and speedy data transfer, this camera creates superbly detailed images and smooth video captures.
  • Included Motic™ Images Plus 3.0 for PC and Mac software enables live and captured images at a maximum quality
  • Edit, measure, and share images or reports with its comprehensive software
  • 12 MP sCMOS camera with USB 3.1 (4000 x 3000 pixels at 25 fps or 2048 x 1080 pixels at 50 fps)
  • Requires: 2 GHz multi-core processor or above, USB 2.0 or USB 3.0 (for high speed transmission), 8 GB RAM, 5 GB (Free Space) hard disk, 1 GB Video RAM on video card
Specifications
Type Microscope Camera
Product Line Moticam S
Includes CS Ring Adaptor, USB 3.1 Cable, Motic 4-dot Calibration Slide, Motic Images Plus 3.0 for PC/OSX/Linux
Sensor Types sCMOS
Size 1/1.7 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov