Learn More
Motic™ Moticam S20 and Analysis Basic Software
Catalog No. 01257469 Shop All Motic Group Products
Change view
Click to view available options
Sensor Types:
sCMOS
Catalog No. Sensor Types
01-257-469 sCMOS
1 options
This item is not returnable. View return policy
Catalog No. 01-257-469 Supplier Motic™ Supplier No. 1100600101401
This item is not returnable. View return policy
The Moticam S20 is the highest resolution model for back-illuminated sensor technology. With speedy data transfer (frame rates up to 50 fps), it creates an optimal imaging environment and makes capturing microscopic images easy.
Motic™'s new C-mount ready sCMOS cameras are the newest members of the Moticam family of professionally designed and engineered cameras. It is easy enough for the microscopy novice, but sophisticated enough for the professional. With back illuminated sensor technology and speedy data transfer, this camera creates superbly detailed images and smooth video captures.
  • Included Motic™ Images Plus 3.0 for PC and Mac software enables live and captured images at a maximum quality
  • Edit, measure, and share images or reports with its comprehensive software
  • 20 MP sCMOS camera with USB 3.1 (5472 x 3648 pixels at 18 fps or 2736 x 1824 pixels at 50 fps)
  • Requires: 2 GHz multi-core processor or above, USB 2.0 or USB 3.0 (for high speed transmission), 8 GB RAM, 5 GB (Free Space) hard disk, 1 GB Video RAM on video card
Specifications
Type Microscope Camera
Product Line Moticam S
Includes CS Ring Adaptor, USB 3.1 Cable, Motic 4-dot Calibration Slide, Motic Images Plus 3.0 for PC/OSX/Linux
Sensor Types sCMOS
Size 1 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov