Learn More
Motic™ Moticam S6 and Analysis Basic Software
Catalog No. 01257466 Shop All Motic Group Products
Change view
Click to view available options
Sensor Types:
sCMOS
Catalog No. Sensor Types
01-257-466 sCMOS
1 options
This item is not returnable. View return policy
Catalog No. 01-257-466 Supplier Motic™ Supplier No. 1100600101111
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Moticam S6 is dedicated to large-sized monitors and printing purposes. Ideal for presentations through a video projector, it also provides images for use in hardcopy with ease.
Motic™'s new C-mount ready sCMOS cameras are the newest members of the Moticam family of professionally designed and engineered cameras. Easy enough for the microscopy novice, but sophisticated enough for the professional. With back illuminated sensor technology and speedy data transfer, this camera creates superbly detailed images and smooth video captures.
  • Included Motic™ Images Plus 3.0 software for PC and Mac enables live and captured images at maximum quality
  • Edit, measure, and share images or reports with Images Plus software
  • 6 MP sCMOS yields 3072 x 2048 pixels at 30 fps or 1536 x 1024 pixels at 50 fps
  • Requires: 2 GHz multi-core processor or above, USB 2.0 or USB 3.0 (for high speed transmission), RAM: 8 GB RAM, 5 GB of space on hard disk, 1 GB video RAM on video card
Specifications
Type Microscope Camera
Product Line Moticam S
Includes CS Ring Adaptor, USB 3.1 Cable, Motic 4-dot Calibration Slide, Motic Images Plus 3.0 for PC/OSX/Linux
Sensor Types sCMOS
Size 1/1.8 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov