Learn More
Motic™ Moticam X5 Plus Camera
Catalog No. 01258039 Shop All Motic Group Products
Change view
Click to view available options
Sensor Types:
CMOS
Catalog No. Sensor Types
01-258-039 CMOS
1 options
This item is not returnable. View return policy
Catalog No. 01-258-039 Supplier Motic™ Supplier No. 1100600101322
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Moticam X5 Plus is a high resolution streaming WiFi camera. By creating its own WiFi signal, simply connect with your Android or iOS device and view, capture, or edit live images from your microscope with our free MotiConnect app.
  • C-mount for lens
  • MotiConnect software is available for free download (with free updates included)
  • Live Display Modes: 1920 x 1080 px, 1280 x 720 px
Specifications
Type WiFi Camera
Includes CS Ring Adapter, Focusable Lens, 30 mm and 38 mm Eyepiece Adapters, USB 2.0 Cable, Motic™ 4-dot Calibration Slide, Macro Tube, Motic™ Images Plus 3.0 PC/OSX/Linux
Sensor Types CMOS
Size 1/3 in.
Frequency (Frame Rate) 30 fps
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov