Learn More
Motic Digital Pathology MoticEasyScan Infinity 100
Catalog No. 01001002 Shop All Motic Group Products
Change view
Click to view available options
For Use With (Equipment):
Glass Microscopy Slides
Catalog No. For Use With (Equipment)
01-001-002 Glass Microscopy Slides
1 options
Catalog No. 01-001-002 Supplier Motic Digital Pathology Supplier No. FSCMDPINF100
Add to Cart
Add to Cart
Request a Quote
Power and speed are key in the MoticEasyScan Infinity. Coming in two models, the MoticEasyScan Infinity 60, with 60-slide capacity, and the MoticEasyScan Infinity 100, with 102-slide capacity. These high-throughput slide scanners offer a flexible configuration for research labs.
Both Infinity models are equipped with three high-quality objectives and a large optical sensor. Intelligent and automated quality control eliminates the need for pre-scan setup while Motic's Smart Tray system slashes hands-on time for technicians and error rates by allowing the real-time tracking and identification of every scanned slide.
  • Large-volume slide scanners with 60- or 102-slide capacity
  • The ideal digital pathology solution for diagnostic and research purposes in high-volume labs
  • Scan more than 1000 slides in an average work week
  • Slides can be added at any time without interruption of scanning processes
  • Exhibits greatly increased efficiency compared with traditional microscopy methods
  • Automatic quality control. LED indicators warn of any potential issues with trays
  • Reduced risk of mechanical failure because no robotic arm is used
  • Provided with a powerful all-in-one PC with a 27 in. QHD-resolution touchscreen
  • Scanning time of as little as 40 seconds per slide
  • Not autoclavable, non-sterile
  • 700 x 398 x 420 mm
Specifications
Product Type 102-Slide Digital Slide Scanner
For Use With (Equipment) Glass Microscopy Slides