Learn More
Motic Digital Pathology MoticFlexScan 1
Catalog No. 01001001 Shop All Motic Group Products
Change view
Click to view available options
For Use With (Equipment):
Flurorescent Microscopy
Catalog No. For Use With (Equipment)
01-001-001 Flurorescent Microscopy
1 options
Catalog No. 01-001-001 Supplier Motic Digital Pathology Supplier No. FSCMDPFLX1
Add to Cart
Add to Cart
Request a Quote
Start lab imaging with MoticFlexScan 1, the affordable and versatile solution for three-color fluorescence scanning and the budget-friendly and easy-to-use desktop-sized solution that offers brightfield and fluorescence whole slide scanning.
  • All-in-one brightfield and fluorescence microscopy: A combination 3-plex and brightfield whole slide scanner that allows for quick analysis right from the desk is an ideal starting point for any project's pre-analysis, optimizing research efficiency and providing valuable insights.
  • Save time on research validation: Cut validation time in half to promptly reach a go or no-go decision on the research.
  • Companion fluorescence system for high-plex research: The low-plex desktop scanner is optimized for preliminary antibody validation, delivering high-resolution whole slide imagery in BF and IF using a high-quality Motic 40x objective lens.
  • Not autoclavable, non-sterile
  • Gray color
  • 202 x 398 x 420 mm
  • For use with fluoroscence applications
  • 1 slide capacity, 1 tray
Specifications
Product Type Fluorescence Whole Slide Imaging Scanner
For Use With (Equipment) Flurorescent Microscopy