Learn More
CPC Scientific H-Phe-Val-Pro-Ile-Phe-Thr-His-Ser-Glu-Leu-Gln-Lys-Ile-Arg-Glu-Lys-Glu-Arg-Asn-Lys-Gly-Gln-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MOTI003B
Encompass
Gastric motor activity-stimulating polypeptide.ONE-LETTER SEQUENCE: FVPIFTHSELQKIREKERNKGQMOLECULAR FORMULA: C120H194N36O34MOLECULAR WEIGHT:2685.08STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [85490-53-5], RESEARCH AREA: Gastrointestinal
Catalog No. 50-210-9822