Learn More
Bioworld Motility Test Agar 500g
Catalog No. 50254335
Encompass_Preferred
Used for the determination of bacterial motility. pH: 7.3, Working Concentration: 22 g/L, Dehydrated, Appearance: beige, homogeneous powder, Prepared Appearance: yellow to tan agar, Soluble in boiling water. Contains (g/L): Pancreatic Digest of Gelatin-10.0g, Beef Extrac- 3.0g , Sodium Chloride- 5.0g, Agar- 4.0g, Appearance Form : Powder. 500g
Catalog No. 50-254-335 Supplier Bioworld Supplier No. 306200531
Used for the determination of bacterial motility. pH: 7.3, Working Concentration: 22 g/L, Dehydrated, Appearance: beige, homogeneous powder, Prepared Appearance: yellow to tan agar, Soluble in boiling water. Contains (g/L): Pancreatic Digest of Gelatin-10.0g, Beef Extrac- 3.0g , Sodium Chloride- 5.0g, Agar- 4.0g, Appearance Form : Powder. 500g