Learn More
Bellco Glass, Inc. The motion alarm monitors the rotation of all rollers. If a belt should break or motor fail in some way, an audible alarm will be produced and the remote alarm jack will be enabled. This alarm will continue until either the problem is fixed or the alarm i
SDP
Catalog No. 501945874
Encompass_Preferred
The motion alarm monitors the rotation of all rollers. If a belt should break or motor fail in some way, an audible alarm will be produced and the remote alarm jack will be enabled. This alarm will continue until either the problem is fixed or the alarm i
Catalog No. 50-194-5874 Supplier Bellco Glass, Inc. Supplier No. 744011111
The motion alarm monitors the rotation of all rollers. If a belt should break or motor fail in some way, an audible alarm will be produced and the remote alarm jack will be enabled. This alarm will continue until either the problem is fixed or the alarm i