Learn More
Medchemexpress LLC HY-P0171 100mg , Motixafortide CAS:664334-36-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0171100MG
Encompass_Preferred
HY-P0171 100mg Motixafortide CAS: 664334-36-5 Motixafortide (BKT140 4-fluorobenzoyl) is a novel CXCR4 antagonist with an IC50 vakue of ~1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0322