Learn More
Medchemexpress LLC Motolimod | 926927-61-9 | 98.1% | 458.60 | 250 MG
SDP
Catalog No. 5000348249
Encompass_Preferred
Motolimod (VTX-2337; VTX-378) is a selective Toll-like receptor 8 (TLR8) agonist with an EC50 of approximately 100 nM. It selectively activates TLR8 among other tested TLRs, stimulating the production of TNFα and IL-12 in PBMCs. This compound is intended for research use only and is not sold to patients.
  • Selective TLR8 agonist with high specificity
  • Stimulates production of TNFα and IL-12 in PBMCs
  • Induces MIP-1β production
  • Increases plasma levels of IL-1β and IL-18 in vivo
  • Supports NK cell activation through IFNγ
  • Evaluated in clinical trials for oncology research
  • Available as a solid, light yellow to brown powder
  • Soluble in DMSO for in vitro applications
Catalog No. 50-003-48249 Supplier Medchemexpress LLC Supplier No. 250MGHY13773
Motolimod (VTX-2337; VTX-378) is a selective Toll-like receptor 8 (TLR8) agonist with an EC50 of approximately 100 nM. It selectively activates TLR8 among other tested TLRs, stimulating the production of TNFα and IL-12 in PBMCs. This compound is intended for research use only and is not sold to patients.
  • Selective TLR8 agonist with high specificity
  • Stimulates production of TNFα and IL-12 in PBMCs
  • Induces MIP-1β production
  • Increases plasma levels of IL-1β and IL-18 in vivo
  • Supports NK cell activation through IFNγ
  • Evaluated in clinical trials for oncology research
  • Available as a solid, light yellow to brown powder
  • Soluble in DMSO for in vitro applications