Learn More
Selleck Chemical LLC Motolimod
SDP
Catalog No. 501364469
Encompass_Preferred
Motolimod is a selective and potent Toll-like receptor 8 (TLR8) agonist with EC50 of 100 nM 50-fold selectivity over TLR7 Phase 2
Catalog No. 50-136-4469 Supplier Selleck Chemical LLC Supplier No. S716125MG
Motolimod is a selective and potent Toll-like receptor 8 (TLR8) agonist with EC50 of 100 nM 50-fold selectivity over TLR7 Phase 2