Learn More
Selleck Chemical LLC Motolimod (VTX-2337) 5mg 926927-61-9
SDP
Catalog No. 501364468
Encompass_Preferred
Motolimod (VTX-2337) is a selective and potent Toll-like receptor 8 (TLR8) agonist with EC50 of 100 nM, > 50-fold selectivity over TLR7. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4468 Supplier Selleck Chemical LLC Supplier No. S71615MG
Motolimod (VTX-2337) is a selective and potent Toll-like receptor 8 (TLR8) agonist with EC50 of 100 nM, > 50-fold selectivity over TLR7. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.