Learn More
Grainger Motor 1 1/10 HP 3 450/1 725 rpm 56Y 230V
Catalog No. 502765085
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B374, B43, B48L, B62, B73, 2HP and 3HP pumps. This motor face measures 7-1/2" diameter and comes with a 1/8 HP 56J frame motor that runs on 115V at 3450 RPM. It has a 1/2" diameter keyed shaft and is totally enclosed and fan cooled. The Century motor face can be installed on existing pump housings to repair older pumps and extend their life. It provides a cost-effective alternative to replacing an entire pump.
Catalog No. 50-276-5085 Supplier Grainger Supplier No. 16U416
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B374, B43, B48L, B62, B73, 2HP and 3HP pumps. This motor face measures 7-1/2" diameter and comes with a 1/8 HP 56J frame motor that runs on 115V at 3450 RPM. It has a 1/2" diameter keyed shaft and is totally enclosed and fan cooled. The Century motor face can be installed on existing pump housings to repair older pumps and extend their life. It provides a cost-effective alternative to replacing an entire pump.