Learn More
Grainger Motor 1 1/2 1/6 HP 3 450/1 725 rpm 230V
Catalog No. 502768589
Encompass_Preferred
The Grainger 1-1/2 HP, 230V, 3-Phase, Open Drip Proof Motor operates at 1725 RPM with a 56C frame. It features a cast iron body with an open drip proof enclosure and pre-lubricated sleeve bearings. This motor is thermally protected and requires a manual reset. It can be used for various industrial applications that require 1-1/2 HP single speed output. NEMA design B with standard starting torque and low starting current.
Catalog No. 50-276-8589 Supplier Grainger Supplier No. 16U425
The Grainger 1-1/2 HP, 230V, 3-Phase, Open Drip Proof Motor operates at 1725 RPM with a 56C frame. It features a cast iron body with an open drip proof enclosure and pre-lubricated sleeve bearings. This motor is thermally protected and requires a manual reset. It can be used for various industrial applications that require 1-1/2 HP single speed output. NEMA design B with standard starting torque and low starting current.