Learn More
Grainger Motor 1 1/2 1/6 HP 3 450/1 725 rpm 230V
Catalog No. 502765397
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, B284F, B284S, and more. This motor face measures 10-1/2" diameter and comes with a 1/2 HP 56J frame motor that runs on 115V with a rotation speed of 1725 RPM. The motor is totally enclosed and fan cooled (TEFC) with a 48-52 frame and 1/2" diameter shaft. It can replace old or worn motor faces to get pool and spa pumps working again. The Century motor face is easy to install and maintain.
Catalog No. 50-276-5397 Supplier Grainger Supplier No. 16U434
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, B284F, B284S, and more. This motor face measures 10-1/2" diameter and comes with a 1/2 HP 56J frame motor that runs on 115V with a rotation speed of 1725 RPM. The motor is totally enclosed and fan cooled (TEFC) with a 48-52 frame and 1/2" diameter shaft. It can replace old or worn motor faces to get pool and spa pumps working again. The Century motor face is easy to install and maintain.