Learn More
Grainger Motor 1 1/2 3/16 HP 3 450/1 725 rpm 230V
Catalog No. 502750645
Encompass_Preferred
The Century Pool and Spa Pump Motor Rigid is a 1.5 HP, 3450 RPM pool pump motor. It has a 48 frame, rigid mount design and is thermally protected for safety. This Century pool pump motor is made of corrosion resistant materials and has a C-face attachment. It can be used as a replacement motor for Hayward, Pentair, and Sta-Rite pool pumps. The motor dimensions are 9-1/2" diameter x 7-3/4" long. It requires 115V power and has a service factor of 1.15. The Century Pool and Spa Pump Motor Rigid provides reliable performance for residential inground swimming pools.
Catalog No. 50-275-0645 Supplier Grainger Supplier No. 14L701
The Century Pool and Spa Pump Motor Rigid is a 1.5 HP, 3450 RPM pool pump motor. It has a 48 frame, rigid mount design and is thermally protected for safety. This Century pool pump motor is made of corrosion resistant materials and has a C-face attachment. It can be used as a replacement motor for Hayward, Pentair, and Sta-Rite pool pumps. The motor dimensions are 9-1/2" diameter x 7-3/4" long. It requires 115V power and has a service factor of 1.15. The Century Pool and Spa Pump Motor Rigid provides reliable performance for residential inground swimming pools.