Learn More
Grainger Motor 1 1/2 HP 3 450 rpm 115/208-230V
Catalog No. 502767652
Encompass_Preferred
The Tripp Lite 16U SmartRack Wall Mount Rack Enclosure Cabinet is a 16U wall-mount rack for storing EIA-standard 19-inch rack equipment in space-constrained areas. Made from heavy-duty steel with a black powder-coated finish, it comes fully assembled for quick installation and can be wall-mounted. The mounting rails adjust to fit equipment up to 20.5 inches deep, and baying tabs allow multiple cabinets to be combined. Features include vented side panels, a removable top cover with cable access holes, and PDU bracket mounting holes. Ideal for organizing rackmount equipment in limited floor space.
Catalog No. 50-276-7652 Supplier Grainger Supplier No. 16U405
The Tripp Lite 16U SmartRack Wall Mount Rack Enclosure Cabinet is a 16U wall-mount rack for storing EIA-standard 19-inch rack equipment in space-constrained areas. Made from heavy-duty steel with a black powder-coated finish, it comes fully assembled for quick installation and can be wall-mounted. The mounting rails adjust to fit equipment up to 20.5 inches deep, and baying tabs allow multiple cabinets to be combined. Features include vented side panels, a removable top cover with cable access holes, and PDU bracket mounting holes. Ideal for organizing rackmount equipment in limited floor space.