Learn More
Grainger Motor 1 1/2 HP 3 450 rpm 115/208-230V
Catalog No. 502764910
Encompass_Preferred
The Tripp Lite 16U SmartRack Wall Mount Rack Enclosure Cabinet is a 16U wall-mountable rack that provides a sturdy way to mount rack equipment in network closets, back offices and other areas with limited floor space. It is constructed with steel for durability and vented sides for proper airflow. The mounting rails adjust in 1/4" increments to accommodate equipment up to 20.5" deep. The front and rear pairs of vertical rails have square mounting holes that allow easy equipment installation. This 16U wall mount rack has a weight capacity of 200 lbs.
Catalog No. 50-276-4910 Supplier Grainger Supplier No. 16U395
The Tripp Lite 16U SmartRack Wall Mount Rack Enclosure Cabinet is a 16U wall-mountable rack that provides a sturdy way to mount rack equipment in network closets, back offices and other areas with limited floor space. It is constructed with steel for durability and vented sides for proper airflow. The mounting rails adjust in 1/4" increments to accommodate equipment up to 20.5" deep. The front and rear pairs of vertical rails have square mounting holes that allow easy equipment installation. This 16U wall mount rack has a weight capacity of 200 lbs.