Learn More
Grainger Motor 1 1/2 HP 3 450 rpm 56J 115/230V
Catalog No. 502768292
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B374, B43, B48L, B62, B73, 2HP and 3HP pumps. The motor face measures approximately 9 inches by 6 inches and has a 1/2 inch diameter shaft. It is designed for 115V applications. This motor face replaces worn or damaged faces to get pool and spa pumps working again.
Catalog No. 50-276-8292 Supplier Grainger Supplier No. 16U478
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B374, B43, B48L, B62, B73, 2HP and 3HP pumps. The motor face measures approximately 9 inches by 6 inches and has a 1/2 inch diameter shaft. It is designed for 115V applications. This motor face replaces worn or damaged faces to get pool and spa pumps working again.