Learn More
Grainger Motor 1 1/2 HP 3 450 rpm 56Y 230V
Catalog No. 502767392
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, N324, and more. This motor face measures 10-1/2 inches by 10-1/2 inches and has a 1/2 inch shaft diameter. It can handle up to 3/4 HP and 3450 RPM. The motor face comes with a thermal protector built in for safety. This is an OEM replacement part made to match the original motor.
Catalog No. 50-276-7392 Supplier Grainger Supplier No. 16U443
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, N324, and more. This motor face measures 10-1/2 inches by 10-1/2 inches and has a 1/2 inch shaft diameter. It can handle up to 3/4 HP and 3450 RPM. The motor face comes with a thermal protector built in for safety. This is an OEM replacement part made to match the original motor.