Learn More
Grainger Motor 1/10 3/4 HP 3 450/1 725 rpm 230V
Catalog No. 502765123
Encompass_Preferred
The Century Square Flange Pool Pump Motor is a 1 HP, 115/230 volt, single phase, 3450 RPM pool pump motor with a 48 frame and a 1.125" diameter shaft. It has a square flange mount and is thermally protected. This Century motor is designed for inground pool pump applications.
Catalog No. 50-276-5123 Supplier Grainger Supplier No. 16U439
The Century Square Flange Pool Pump Motor is a 1 HP, 115/230 volt, single phase, 3450 RPM pool pump motor with a 48 frame and a 1.125" diameter shaft. It has a square flange mount and is thermally protected. This Century motor is designed for inground pool pump applications.