Learn More
Grainger Motor 1/10 3/4 HP 3 450/1 725 rpm 115V
Catalog No. 502767251
Encompass_Preferred
The Grainger 1/10 HP 115V 1725 RPM Single Phase Open Drip Proof Motor is a 3450 RPM motor with a 1/10 horsepower rating and 115 volt power requirement. It has a 48Y frame, operates on 60 Hz frequency, and features a sleeve bearing. This motor is thermally protected, CSA certified, and RoHS compliant. With a standard NEMA mounting configuration, this motor is suitable for a variety of light-duty industrial, commercial, and residential applications.
Catalog No. 50-276-7251 Supplier Grainger Supplier No. 16U437
The Grainger 1/10 HP 115V 1725 RPM Single Phase Open Drip Proof Motor is a 3450 RPM motor with a 1/10 horsepower rating and 115 volt power requirement. It has a 48Y frame, operates on 60 Hz frequency, and features a sleeve bearing. This motor is thermally protected, CSA certified, and RoHS compliant. With a standard NEMA mounting configuration, this motor is suitable for a variety of light-duty industrial, commercial, and residential applications.