Learn More
Grainger Motor 1/2 HP 3 450 rpm 56Y 115/208-230V
Catalog No. 502765135
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, B284F, B284S, and more. This motor face measures 10-1/2" diameter and comes with a 1/2 HP 56J frame motor that runs on 115V with a rotation speed of 1725 RPM. The motor is totally enclosed and fan cooled (TEFC) with a 48-52 frame and 1/2" diameter C-face. It can replace old or worn motor faces to get pool and spa pumps working again.
Catalog No. 50-276-5135 Supplier Grainger Supplier No. 16U451
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, B284F, B284S, and more. This motor face measures 10-1/2" diameter and comes with a 1/2 HP 56J frame motor that runs on 115V with a rotation speed of 1725 RPM. The motor is totally enclosed and fan cooled (TEFC) with a 48-52 frame and 1/2" diameter C-face. It can replace old or worn motor faces to get pool and spa pumps working again.