Learn More
Grainger Motor 1/2 HP 3 450 rpm 56Y 115/208-230V
Catalog No. 502765109
Encompass_Preferred
The Century Square Flange Pool Pump Motor is a 1 HP, 115V, single phase, 3450 RPM pool pump motor with a 48 frame and a square flange. It has a NEMA standard C-face mounting and is constructed with a heavy-duty ball bearing design. The motor is totally enclosed and fan cooled (TEFC) with a Class F insulation and automatic thermal overload protection. It can be used as a replacement motor for most major pool pump brands. The motor dimensions are approximately 9-1/2" long by 5-3/4" wide and it weighs 15 lbs.
Catalog No. 50-276-5109 Supplier Grainger Supplier No. 16U409
The Century Square Flange Pool Pump Motor is a 1 HP, 115V, single phase, 3450 RPM pool pump motor with a 48 frame and a square flange. It has a NEMA standard C-face mounting and is constructed with a heavy-duty ball bearing design. The motor is totally enclosed and fan cooled (TEFC) with a Class F insulation and automatic thermal overload protection. It can be used as a replacement motor for most major pool pump brands. The motor dimensions are approximately 9-1/2" long by 5-3/4" wide and it weighs 15 lbs.