Learn More
Grainger Motor 1/3 2 HP 3 450/1 725 rpm 56Y 230V
Catalog No. 502767332
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B374, B43, B48L, B62, B73, 2HP and 3HP pumps. The motor face measures 7-1/2 inches in diameter and comes with a 1 year manufacturer's warranty. This Century motor face is designed for inground pools and spas and is easy to install.
Catalog No. 50-276-7332 Supplier Grainger Supplier No. 16U436
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B374, B43, B48L, B62, B73, 2HP and 3HP pumps. The motor face measures 7-1/2 inches in diameter and comes with a 1 year manufacturer's warranty. This Century motor face is designed for inground pools and spas and is easy to install.