Learn More
Grainger Motor 1/3 3 HP 3 450/1 725 rpm 56Z 230V
Catalog No. 502767283
Encompass_Preferred
The Century Pool and Spa Pump Motor Rigid is a 1 HP, 115V, single phase pool pump motor with a rigid shaft. It has a 48 frame size and is thermally protected. This Century pool pump motor is designed for inground pools and spas and is made of corrosion resistant construction. It features permanently lubricated ball bearings and is CSA listed. The motor has a rotation of 1725 RPM and is for use with Century, Purex/Pleatco, Unicel, and other major filter brands. This rigid shaft pool pump motor is ideal for replacing worn out or defective pump motors.
Catalog No. 50-276-7283 Supplier Grainger Supplier No. 16U427
The Century Pool and Spa Pump Motor Rigid is a 1 HP, 115V, single phase pool pump motor with a rigid shaft. It has a 48 frame size and is thermally protected. This Century pool pump motor is designed for inground pools and spas and is made of corrosion resistant construction. It features permanently lubricated ball bearings and is CSA listed. The motor has a rotation of 1725 RPM and is for use with Century, Purex/Pleatco, Unicel, and other major filter brands. This rigid shaft pool pump motor is ideal for replacing worn out or defective pump motors.