Learn More
Grainger Motor 1/4 2 HP 3 450/1 725 rpm 56J 230V
Catalog No. 502765767
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, B284F, and more. This motor face measures 10-1/2 inches by 10-1/2 inches and has a 1/2 inch shaft diameter. It can handle up to 3/4 HP and has a 56J frame size. The motor operates on 115V with a rotation speed of 1725 RPM. This is an OEM replacement part made to match the original motor for a proper fit and performance.
Catalog No. 50-276-5767 Supplier Grainger Supplier No. 16U463
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, B284F, and more. This motor face measures 10-1/2 inches by 10-1/2 inches and has a 1/2 inch shaft diameter. It can handle up to 3/4 HP and has a 56J frame size. The motor operates on 115V with a rotation speed of 1725 RPM. This is an OEM replacement part made to match the original motor for a proper fit and performance.