Learn More
Grainger Motor 1/4 2 HP 3 450/1 725 rpm 56Z 230V
Catalog No. 502765917
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, N324, N236, and more. This motor face measures 10-1/2" diameter x 7-3/4" high and comes with a 1 year limited warranty. It is designed for use in inground pools and spas and features a 56J frame, 1/2 HP, 3450 RPM, 48Y frame, and 1 phase. The motor face attaches to the back of the pump and drives the impeller.
Catalog No. 50-276-5917 Supplier Grainger Supplier No. 16U424
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, N324, N236, and more. This motor face measures 10-1/2" diameter x 7-3/4" high and comes with a 1 year limited warranty. It is designed for use in inground pools and spas and features a 56J frame, 1/2 HP, 3450 RPM, 48Y frame, and 1 phase. The motor face attaches to the back of the pump and drives the impeller.