Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Motor 1/4 HP 1725 rpm 56C 230/460V

Catalog No. 502755865
Encompass_Preferred

Motor 1/4 HP 1725 rpm 56C 230/460V

Catalog No. 50-275-5865 Supplier Grainger Supplier No. 15G104
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Motor 1/4 HP 1725 rpm 56C 230/460V

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.